Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMCN2
Entrez Gene ID
256158
Gene Name
hemicentin 2
Gene Aliases
-
Chromosome Location
Chr.9: 130265742 - 130434123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291815.1 NP_001278744.1 95 - 96 14653 92
XM_011518465.2 XP_011516767.1 94 - 95 14667 92
XM_011518466.2 XP_011516768.1 94 - 95 14658 92
XM_011518467.2 XP_011516769.1 94 - 95 14613 92
XM_017014585.1 XP_016870074.1 74 - 75 11459 92
XM_017014586.1 XP_016870075.1 49 - 50 7387 92
GenBank mRNA AK074396.1 - 1 - 2 330 92

View Full Product Details