The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033386.3 | NP_203744.1 | 2 - 3 | 469 | 62 |
XM_005261791.3 | XP_005261848.1 | 2 - 3 | 282 | 62 | |
XM_005261792.3 | XP_005261849.1 | 2 - 3 | 282 | 62 | |
XM_011530471.2 | XP_011528773.1 | 2 - 3 | 282 | 62 | |
XM_011530472.2 | XP_011528774.1 | 2 - 3 | 282 | 62 | |
XM_011530473.2 | XP_011528775.1 | 2 - 3 | 282 | 62 | |
XM_011530474.2 | XP_011528776.1 | 2 - 3 | 693 | 62 | |
XM_011530475.2 | XP_011528777.1 | 2 - 3 | 282 | 62 | |
XM_011530476.2 | XP_011528778.1 | 2 - 3 | 282 | 62 | |
GenBank mRNA | AJ496196.1 | - | 2 - 3 | 261 | 62 |
AL834373.1 | - | 2 - 3 | 123 | 62 | |
CB997694.1 | - | 2 - 3 | 347 | 62 | |
CR456437.1 | - | 2 - 3 | 200 | 62 |