The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033386.3 | NP_203744.1 | 3 - 4 | 609 | 73 |
XM_005261791.3 | XP_005261848.1 | 3 - 4 | 422 | 73 | |
XM_005261792.3 | XP_005261849.1 | 3 - 4 | 422 | 73 | |
XM_011530471.2 | XP_011528773.1 | 3 - 4 | 422 | 73 | |
XM_011530472.2 | XP_011528774.1 | 3 - 4 | 422 | 73 | |
XM_011530473.2 | XP_011528775.1 | 3 - 4 | 422 | 73 | |
XM_011530474.2 | XP_011528776.1 | 3 - 4 | 833 | 73 | |
XM_011530475.2 | XP_011528777.1 | 3 - 4 | 422 | 73 | |
XM_011530476.2 | XP_011528778.1 | 3 - 4 | 422 | 73 | |
GenBank mRNA | AB051455.1 | - | 1 - 2 | 119 | 73 |
AJ496196.1 | - | 3 - 4 | 401 | 73 | |
CR456437.1 | - | 3 - 4 | 340 | 73 |