The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033386.3 | NP_203744.1 | 8 - 9 | 1743 | 118 |
XM_005261791.3 | XP_005261848.1 | 8 - 9 | 1556 | 118 | |
XM_005261792.3 | XP_005261849.1 | 8 - 9 | 1556 | 118 | |
GenBank mRNA | AB051455.1 | - | 6 - 7 | 1253 | 118 |
AJ496196.1 | - | 8 - 9 | 1535 | 118 | |
AK056603.1 | - | 2 - 3 | 439 | 118 | |
AL833860.1 | - | 3 - 4 | 852 | 118 | |
CR456437.1 | - | 8 - 9 | 1474 | 118 |