The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153712.4 | NP_714923.1 | 2 - 3 | 420 | 104 |
XM_005263599.3 | XP_005263656.1 | 2 - 3 | 459 | 104 | |
XM_011510665.2 | XP_011508967.1 | 2 - 3 | 459 | 104 | |
GenBank mRNA | AB071393.2 | - | 2 - 3 | 265 | 104 |
BC036819.1 | - | 2 - 3 | 432 | 104 |