The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291970.1 | NP_001278899.1 | 3 - 4 | 577 | 84 |
NM_006502.2 | NP_006493.1 | 3 - 4 | 577 | 84 | |
XM_005249186.3 | XP_005249243.1 | 2 - 3 | 430 | 84 | |
GenBank mRNA | AB024313.1 | - | 3 - 4 | 510 | 84 |
AF158185.1 | - | 3 - 4 | 438 | 84 | |
AK023893.1 | - | 3 - 4 | 568 | 84 | |
AK294430.1 | - | 3 - 4 | 558 | 84 | |
BC015742.1 | - | 3 - 4 | 421 | 84 |