Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THEMIS2
Entrez Gene ID
9473
Gene Name
thymocyte selection associated family member 2
Gene Aliases
C1orf38, ICB-1
Chromosome Location
Chr.1: 27872531 - 27886685 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105556.2 NP_001099026.1 4 - 5 1743 92
NM_001286113.1 NP_001273042.1 5 - 6 1356 92
NM_001286115.1 NP_001273044.1 4 - 5 1155 92
XM_005246041.3 XP_005246098.1 3 - 4 1344 92
XM_006711050.1 XP_006711113.1 5 - 6 1302 92
XM_011542445.2 XP_011540747.1 3 - 4 1614 92
GenBank mRNA AB050854.1 - 3 - 4 960 92
AF044896.1 - 2 - 3 1397 92
AK303090.1 - 5 - 6 1318 92
AK303141.1 - 4 - 5 1151 92
BI908174.1 - 1 - 2 265 92

View Full Product Details