The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002514.3 | NP_002505.1 | 1 - 2 | 305 | 150 |
GenBank mRNA | AK294289.1 | - | 1 - 2 | 177 | 150 |
AK312355.1 | - | 1 - 2 | 171 | 150 | |
AY082381.1 | - | 1 - 2 | 156 | 150 | |
BC015028.1 | - | 1 - 2 | 151 | 150 | |
CR457432.1 | - | 1 - 2 | 84 | 150 | |
DA162276.1 | - | 1 - 2 | 311 | 150 | |
X96584.1 | - | 1 - 2 | 156 | 150 |