The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019116.2 | NP_061989.2 | 1 - 2 | 230 | 85 |
XM_011545894.2 | XP_011544196.1 | 1 - 2 | 236 | 85 | |
XM_017023459.1 | XP_016878948.1 | 1 - 2 | 236 | 85 | |
XM_017023460.1 | XP_016878949.1 | 1 - 2 | 236 | 85 |