The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002755.3 | NP_002746.1 | 6 - 7 | 1170 | 80 |
XM_011521783.2 | XP_011520085.1 | 6 - 7 | 827 | 80 | |
XM_017022411.1 | XP_016877900.1 | 5 - 6 | 1043 | 80 | |
XM_017022412.1 | XP_016877901.1 | 5 - 6 | 629 | 80 | |
XM_017022413.1 | XP_016877902.1 | 2 - 3 | 1238 | 80 | |
GenBank mRNA | AK291500.1 | - | 6 - 7 | 1131 | 80 |
AK294322.1 | - | 6 - 7 | 681 | 80 | |
AK312356.1 | - | 6 - 7 | 1077 | 80 | |
BC137459.1 | - | 6 - 7 | 758 | 80 | |
BC139729.1 | - | 6 - 7 | 754 | 80 | |
L05624.1 | - | 6 - 7 | 728 | 80 | |
L11284.1 | - | 6 - 7 | 767 | 80 |