Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNS
Entrez Gene ID
2799
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Gene Aliases
G6S
Chromosome Location
Chr.12: 64713442 - 64759446 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002076.3 NP_002067.1 9 - 10 1265 90
GenBank mRNA AK223484.1 - 9 - 10 1222 90
AK291771.1 - 9 - 10 1201 90
AK300350.1 - 9 - 10 1062 90
AK302443.1 - 8 - 9 1163 90
AK304158.1 - 10 - 11 1321 90
AK308094.1 - 10 - 11 1179 90
BC012482.1 - 9 - 10 1226 90
BX537363.1 - 8 - 9 1017 90
KU178097.1 - 9 - 10 1095 90
Z12173.1 - 9 - 10 1182 90

View Full Product Details