The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042424.2 | NP_001035889.1 | 12 - 13 | 2513 | 89 |
NM_133330.2 | NP_579877.1 | 14 - 15 | 2831 | 89 | |
NM_133331.2 | NP_579878.1 | 13 - 14 | 2651 | 89 | |
NM_133335.3 | NP_579890.1 | 12 - 13 | 2476 | 89 | |
XM_005248001.3 | XP_005248058.1 | 12 - 13 | 2456 | 89 | |
XM_005248002.2 | XP_005248059.1 | 10 - 11 | 2345 | 89 | |
XM_011513557.2 | XP_011511859.1 | 12 - 13 | 2392 | 89 | |
XM_017008587.1 | XP_016864076.1 | 3 - 4 | 359 | 89 | |
XM_017008588.1 | XP_016864077.1 | 2 - 3 | 1089 | 89 | |
GenBank mRNA | AB029013.1 | - | 2 - 3 | 903 | 89 |
AF071593.1 | - | 11 - 12 | 2366 | 89 | |
AF083386.1 | - | 14 - 15 | 2831 | 89 | |
AF083387.1 | - | 13 - 14 | 2651 | 89 | |
AF083388.1 | - | 12 - 13 | 2475 | 89 | |
AF083389.1 | - | 15 - 16 | 3058 | 89 | |
AF083390.1 | - | 15 - 16 | 2925 | 89 | |
AF330040.1 | - | 4 - 5 | 954 | 89 | |
AJ007042.1 | - | 12 - 13 | 3099 | 89 | |
BC144464.1 | - | 3 - 4 | 363 | 89 | |
BC152412.1 | - | 2 - 3 | 938 | 89 |