Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTRK3
Entrez Gene ID
4916
Gene Name
neurotrophic receptor tyrosine kinase 3
Gene Aliases
GP145-TrkC, TRKC, gp145(trkC)
Chromosome Location
Chr.15: 87859749 - 88256796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007156.2 NP_001007157.1 7 - 8 926 60
NM_001012338.2 NP_001012338.1 7 - 8 926 60
NM_001243101.1 NP_001230030.1 7 - 8 926 60
NM_001320134.1 NP_001307063.1 5 - 6 636 60
NM_001320135.1 NP_001307064.1 5 - 6 527 60
NM_002530.3 NP_002521.2 7 - 8 926 60
XM_006720543.3 XP_006720606.1 5 - 6 1235 60
XM_006720544.3 XP_006720607.1 5 - 6 1235 60
XM_006720545.3 XP_006720608.1 5 - 6 1235 60
XM_006720548.3 XP_006720611.1 5 - 6 1235 60
XM_006720549.3 XP_006720612.1 5 - 6 1235 60
XM_006720550.3 XP_006720613.1 5 - 6 1235 60
XM_011521637.2 XP_011519939.1 5 - 6 1235 60
XM_011521638.2 XP_011519940.1 5 - 6 1235 60
XM_017022240.1 XP_016877729.1 6 - 7 1106 60
XM_017022241.1 XP_016877730.1 5 - 6 591 60
XM_017022242.1 XP_016877731.1 5 - 6 1235 60
XM_017022243.1 XP_016877732.1 5 - 6 587 60
XM_017022244.1 XP_016877733.1 5 - 6 489 60
XM_017022245.1 XP_016877734.1 6 - 7 544 60
XM_017022246.1 XP_016877735.1 5 - 6 645 60
XM_017022247.1 XP_016877736.1 5 - 6 1235 60
XM_017022248.1 XP_016877737.1 5 - 6 1235 60
XM_017022249.1 XP_016877738.1 5 - 6 1235 60
XM_017022250.1 XP_016877739.1 5 - 6 1235 60
XM_017022253.1 XP_016877742.1 5 - 6 1235 60
XM_017022254.1 XP_016877743.1 5 - 6 1235 60
GenBank mRNA AK297160.1 - 5 - 6 644 60
AK302504.1 - 5 - 6 486 60
AK309695.1 - 5 - 6 646 60
AK313646.1 - 5 - 6 645 60
BC013693.1 - 5 - 6 625 60
BC128249.1 - 1 - 2 82 60
BT007291.1 - 5 - 6 620 60
HY003065.1 - 5 - 6 527 60
KJ534904.1 - 5 - 6 620 60
KJ534905.1 - 5 - 6 620 60
KJ534906.1 - 5 - 6 620 60
KJ535052.1 - 5 - 6 620 60
S76475.1 - 5 - 6 635 60
S76476.1 - 5 - 6 635 60
U05012.1 - 6 - 7 775 60

View Full Product Details