The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039775.3 | NP_001034864.2 | 16 - 17 | 4607 | 59 |
XM_005245918.2 | XP_005245975.1 | 16 - 17 | 4593 | 59 | |
XM_011541672.1 | XP_011539974.1 | 15 - 16 | 4557 | 59 | |
XM_011541673.2 | XP_011539975.1 | 16 - 17 | 4832 | 59 | |
GenBank mRNA | AK095339.1 | - | 15 - 16 | 1651 | 59 |
BC136870.1 | - | 12 - 13 | 1349 | 59 |