Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLPTM1L
Entrez Gene ID
81037
Gene Name
CLPTM1 like
Gene Aliases
CRR9
Chromosome Location
Chr.5: 1317744 - 1345070 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030782.4 NP_110409.2 13 - 14 1547 67
XM_011514144.2 XP_011512446.1 13 - 14 1511 67
GenBank mRNA AB045223.1 - 13 - 14 1304 67
AK027306.1 - 13 - 14 1361 67
AK128462.1 - 4 - 5 2740 67
AL832953.1 - 3 - 4 149 67
BC016399.1 - 10 - 11 825 67
BC025305.1 - 13 - 14 1365 67

View Full Product Details