The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030782.4 | NP_110409.2 | 16 - 17 | 1759 | 66 |
XM_011514144.2 | XP_011512446.1 | 16 - 17 | 1723 | 66 | |
GenBank mRNA | AB045223.1 | - | 16 - 17 | 1516 | 66 |
AK027306.1 | - | 16 - 17 | 1573 | 66 | |
AK128462.1 | - | 7 - 8 | 2952 | 66 | |
AL832953.1 | - | 6 - 7 | 361 | 66 | |
BC016399.1 | - | 13 - 14 | 1037 | 66 | |
BC025305.1 | - | 16 - 17 | 1577 | 66 |