The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174089.1 | NP_001167560.1 | 4 - 5 | 389 | 79 |
NM_001174090.1 | NP_001167561.1 | 4 - 5 | 539 | 79 | |
NM_032034.3 | NP_114423.1 | 3 - 4 | 381 | 79 | |
NR_135000.1 | 4 - 5 | 453 | 79 | ||
XM_005260856.4 | XP_005260913.1 | 4 - 5 | 877 | 79 | |
XM_005260857.1 | XP_005260914.1 | 4 - 5 | 408 | 79 | |
XM_011529383.2 | XP_011527685.1 | 4 - 5 | 1984 | 79 | |
XM_011529384.1 | XP_011527686.1 | 4 - 5 | 626 | 79 | |
XM_011529385.1 | XP_011527687.1 | 4 - 5 | 453 | 79 | |
XM_017028093.1 | XP_016883582.1 | 4 - 5 | 877 | 79 | |
XM_017028094.1 | XP_016883583.1 | 4 - 5 | 261 | 79 | |
XM_017028095.1 | XP_016883584.1 | 4 - 5 | 732 | 79 | |
XM_017028096.1 | XP_016883585.1 | 5 - 6 | 638 | 79 | |
XM_017028097.1 | XP_016883586.1 | 4 - 5 | 877 | 79 | |
GenBank mRNA | AF336127.1 | - | 3 - 4 | 381 | 79 |
AK091157.1 | - | 4 - 5 | 453 | 79 | |
AK296508.1 | - | 4 - 5 | 539 | 79 | |
AK296760.1 | - | 4 - 5 | 338 | 79 | |
BC110540.1 | - | 3 - 4 | 354 | 79 | |
BC110541.1 | - | 3 - 4 | 354 | 79 | |
DA630556.1 | - | 4 - 5 | 453 | 79 | |
DC341640.1 | - | 4 - 5 | 389 | 79 |