The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022760.4 | NP_073597.2 | 1 - 2 | 493 | 78 |
XM_005260805.2 | XP_005260862.1 | 1 - 2 | 426 | 78 | |
XM_005260806.3 | XP_005260863.1 | 1 - 2 | 426 | 78 | |
GenBank mRNA | AK056353.1 | - | 1 - 2 | 477 | 78 |
AK293638.1 | - | 1 - 2 | 266 | 78 | |
BC008864.2 | - | - | 391 | 78 | |
BC037240.1 | - | 1 - 2 | 99 | 78 | |
BC051816.1 | - | 1 - 2 | 281 | 78 | |
DC421675.1 | - | 1 - 2 | 493 | 78 |