The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271168.1 | NP_001258097.1 | 2 - 3 | 520 | 65 |
NM_022760.4 | NP_073597.2 | 2 - 3 | 634 | 65 | |
XM_005260804.2 | XP_005260861.1 | 2 - 3 | 520 | 65 | |
XM_005260805.2 | XP_005260862.1 | 2 - 3 | 567 | 65 | |
XM_005260806.3 | XP_005260863.1 | 2 - 3 | 567 | 65 | |
GenBank mRNA | AK026029.1 | - | 2 - 3 | 428 | 65 |
AK056353.1 | - | 2 - 3 | 618 | 65 | |
AK293638.1 | - | 2 - 3 | 407 | 65 | |
BC008864.2 | - | - | 532 | 65 | |
BC014247.2 | - | 2 - 3 | 421 | 65 | |
BC037240.1 | - | 2 - 3 | 240 | 65 | |
BC051816.1 | - | 2 - 3 | 422 | 65 | |
BE302577.1 | - | 2 - 3 | 467 | 65 |