Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THEMIS2
Entrez Gene ID
9473
Gene Name
thymocyte selection associated family member 2
Gene Aliases
C1orf38, ICB-1
Chromosome Location
Chr.1: 27872531 - 27886685 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039477.2 NP_001034566.1 1 - 2 126 116
NM_001105556.2 NP_001099026.1 1 - 2 126 116
NM_001286113.1 NP_001273042.1 1 - 2 126 116
NM_001286115.1 NP_001273044.1 1 - 2 126 116
NM_004848.3 NP_004839.2 1 - 2 126 116
XM_005246041.3 XP_005246098.1 1 - 2 138 116
XM_006711050.1 XP_006711113.1 1 - 2 120 116
GenBank mRNA AK303090.1 - 1 - 2 88 116
AK303141.1 - 1 - 2 122 116
AL554503.3 - 1 - 2 90 116
BC031655.1 - 1 - 2 94 116
BC132692.1 - 1 - 2 125 116
BC133049.1 - 1 - 2 125 116
BG716895.1 - 1 - 2 119 116
BM146655.1 - 1 - 2 126 116
CR749321.1 - 1 - 2 127 116

View Full Product Details