Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLIPR1L1
Entrez Gene ID
256710
Gene Name
GLI pathogenesis related 1 like 1
Gene Aliases
ALKN2972, PRO7434
Chromosome Location
Chr.12: 75331834 - 75370390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304964.1 NP_001291893.1 4 - 5 701 97
GenBank mRNA AY358731.1 - 4 - 5 701 97

View Full Product Details