The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110514.1 | NP_001103984.1 | 4 - 5 | 618 | 55 |
XM_006723600.1 | XP_006723663.1 | 4 - 5 | 413 | 55 | |
XM_017027983.1 | XP_016883472.1 | 5 - 6 | 510 | 55 | |
XM_017027984.1 | XP_016883473.1 | 5 - 6 | 616 | 55 | |
GenBank mRNA | AB037863.1 | - | 4 - 5 | 612 | 55 |