Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCDC5
Entrez Gene ID
100506627
Gene Name
doublecortin domain containing 5
Gene Aliases
-
Chromosome Location
Chr.11: 30863603 - 30992686 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB040926.1 - 3 - 4 674 110

View Full Product Details