The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020869.3 | NP_065920.2 | 7 - 8 | 1091 | 88 |
GenBank mRNA | BX647348.1 | - | 7 - 8 | 1091 | 88 |