The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020869.3 | NP_065920.2 | 19 - 20 | 3031 | 112 |
GenBank mRNA | AB040926.1 | - | 10 - 11 | 3043 | 112 |
BX117620.1 | - | 3 - 4 | 356 | 112 | |
BX647348.1 | - | 18 - 19 | 4307 | 112 |