The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243101.1 | NP_001230030.1 | 10 - 11 | 1511 | 103 |
XM_006720544.3 | XP_006720607.1 | 8 - 9 | 1820 | 103 | |
XM_006720550.3 | XP_006720613.1 | 8 - 9 | 1820 | 103 | |
XM_017022247.1 | XP_016877736.1 | 8 - 9 | 1820 | 103 | |
XM_017022248.1 | XP_016877737.1 | 8 - 9 | 1820 | 103 | |
GenBank mRNA | AK297160.1 | - | 8 - 9 | 1229 | 103 |
AK309695.1 | - | 8 - 9 | 1231 | 103 | |
AK313646.1 | - | 8 - 9 | 1230 | 103 | |
KJ534906.1 | - | 8 - 9 | 1205 | 103 | |
KJ535052.1 | - | 8 - 9 | 1205 | 103 |