The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020904.2 | NP_065955.2 | 16 - 17 | 2289 | 132 |
XM_005259107.2 | XP_005259164.1 | 15 - 16 | 2209 | 132 | |
XM_006723301.2 | XP_006723364.1 | 15 - 16 | 2209 | 132 | |
XM_011527157.1 | XP_011525459.1 | 16 - 17 | 2296 | 132 | |
XM_011527158.2 | XP_011525460.1 | 15 - 16 | 2135 | 132 | |
XM_011527159.1 | XP_011525461.1 | 16 - 17 | 2309 | 132 | |
XM_011527160.1 | XP_011525462.1 | 16 - 17 | 2296 | 132 | |
XM_011527162.2 | XP_011525464.1 | 9 - 10 | 905 | 132 | |
XM_017027037.1 | XP_016882526.1 | 13 - 14 | 1573 | 132 | |
GenBank mRNA | AB208908.1 | - | 14 - 15 | 3781 | 132 |
BC024157.2 | - | 16 - 17 | 2269 | 132 | |
BC033832.1 | - | 8 - 9 | 739 | 132 |