The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161354.1 | NP_001154826.1 | 4 - 5 | 807 | 66 |
NM_020904.2 | NP_065955.2 | 4 - 5 | 807 | 66 | |
XM_005259107.2 | XP_005259164.1 | 4 - 5 | 802 | 66 | |
XM_006723301.2 | XP_006723364.1 | 4 - 5 | 802 | 66 | |
XM_011527157.1 | XP_011525459.1 | 4 - 5 | 802 | 66 | |
XM_011527158.2 | XP_011525460.1 | 3 - 4 | 641 | 66 | |
XM_011527160.1 | XP_011525462.1 | 4 - 5 | 802 | 66 | |
GenBank mRNA | AY007233.1 | - | 4 - 5 | 781 | 66 |
BC024157.2 | - | 4 - 5 | 787 | 66 | |
BC064601.1 | - | 4 - 5 | 634 | 66 |