The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105563.1 | NP_001099033.1 | 9 - 10 | 1555 | 53 |
NM_001105564.1 | NP_001099034.1 | 9 - 10 | 1663 | 53 | |
NM_019052.3 | NP_061925.2 | 9 - 10 | 1330 | 53 | |
XM_011514702.2 | XP_011513004.1 | 9 - 10 | 1589 | 53 | |
XM_011514703.1 | XP_011513005.1 | 10 - 11 | 1402 | 53 | |
XM_011514704.2 | XP_011513006.1 | 9 - 10 | 1191 | 53 | |
XM_011514705.1 | XP_011513007.1 | 9 - 10 | 1300 | 53 | |
XM_017010961.1 | XP_016866450.1 | 9 - 10 | 1697 | 53 | |
XM_017010962.1 | XP_016866451.1 | 10 - 11 | 1449 | 53 | |
XM_017010963.1 | XP_016866452.1 | 10 - 11 | 1563 | 53 | |
XM_017010964.1 | XP_016866453.1 | 9 - 10 | 1327 | 53 | |
XM_017010965.1 | XP_016866454.1 | 9 - 10 | 1389 | 53 | |
XM_017010966.1 | XP_016866455.1 | 9 - 10 | 1410 | 53 | |
XM_017010967.1 | XP_016866456.1 | 9 - 10 | 1513 | 53 | |
XM_017010968.1 | XP_016866457.1 | 9 - 10 | 1282 | 53 | |
XM_017010969.1 | XP_016866458.1 | 9 - 10 | 1189 | 53 | |
XM_017010970.1 | XP_016866459.1 | 8 - 9 | 975 | 53 | |
GenBank mRNA | AB029331.1 | - | 7 - 8 | 1151 | 53 |
AB112474.1 | - | 9 - 10 | 1286 | 53 | |
AB112475.1 | - | 9 - 10 | 1637 | 53 | |
AF216493.1 | - | 7 - 8 | 1151 | 53 | |
AK000204.1 | - | 9 - 10 | 1286 | 53 | |
AK000217.1 | - | 9 - 10 | 1287 | 53 | |
AK289446.1 | - | 9 - 10 | 1278 | 53 | |
AK295494.1 | - | 9 - 10 | 1350 | 53 | |
AK310626.1 | - | 9 - 10 | 1348 | 53 | |
AY029160.1 | - | 8 - 9 | 1207 | 53 | |
BC110534.1 | - | 9 - 10 | 1373 | 53 | |
BC110535.1 | - | 9 - 10 | 1486 | 53 | |
CR457227.1 | - | 8 - 9 | 1207 | 53 |