Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NADK2
Entrez Gene ID
133686
Gene Name
NAD kinase 2, mitochondrial
Gene Aliases
C5orf33, DECRD, MNADK, NADKD1
Chromosome Location
Chr.5: 36192589 - 36242279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085411.2 NP_001078880.1 5 - 6 643 89
NM_001287340.1 NP_001274269.1 5 - 6 376 89
NM_001287341.1 NP_001274270.1 5 - 6 527 89
NM_153013.4 NP_694558.1 5 - 6 527 89
XM_005248241.4 XP_005248298.1 5 - 6 739 89
GenBank mRNA AK055158.1 - 5 - 6 376 89
AK304009.1 - 5 - 6 528 89
AY360463.1 - 2 - 3 154 89
BC062567.1 - 5 - 6 390 89

View Full Product Details