The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015880.1 | NP_001015880.1 | 11 - 12 | 1752 | 100 |
NM_004670.3 | NP_004661.2 | 10 - 11 | 1737 | 100 | |
GenBank mRNA | AF074331.1 | - | 10 - 11 | 1537 | 100 |
AF091242.1 | - | 10 - 11 | 1554 | 100 | |
AF150754.2 | - | 11 - 12 | 1569 | 100 | |
AF173365.1 | - | 11 - 12 | 1537 | 100 | |
AF313907.1 | - | 10 - 11 | 1474 | 100 | |
AK301453.1 | - | 10 - 11 | 1524 | 100 | |
AK312556.1 | - | 10 - 11 | 1578 | 100 | |
BC009894.2 | - | 10 - 11 | 1585 | 100 |