The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032211.6 | NP_115587.6 | 10 - 11 | 1746 | 64 |
XM_005270216.1 | XP_005270273.1 | 10 - 11 | 1722 | 64 | |
XM_006718020.1 | XP_006718083.1 | 11 - 12 | 1912 | 64 | |
GenBank mRNA | AF338441.1 | - | 9 - 10 | 1595 | 64 |
AK025542.1 | - | 1 - 2 | 124 | 64 | |
AK172781.1 | - | 10 - 11 | 1689 | 64 | |
AY036093.1 | - | 9 - 10 | 1601 | 64 | |
BC007522.1 | - | 3 - 4 | 462 | 64 | |
BC013153.1 | - | 10 - 11 | 1746 | 64 | |
BC035470.1 | - | 10 - 11 | 1746 | 64 |