The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276713.1 | NP_001263642.1 | 7 - 8 | 956 | 71 |
XM_011543618.2 | XP_011541920.1 | 7 - 8 | 1143 | 71 | |
XM_011543623.2 | XP_011541925.1 | 3 - 4 | 708 | 71 | |
XM_017009814.1 | XP_016865303.1 | 6 - 7 | 1248 | 71 | |
XM_017009815.1 | XP_016865304.1 | 6 - 7 | 1248 | 71 |