The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276713.1 | NP_001263642.1 | 9 - 10 | 1158 | 97 |
XM_011543618.2 | XP_011541920.1 | 9 - 10 | 1345 | 97 | |
XM_011543623.2 | XP_011541925.1 | 5 - 6 | 910 | 97 | |
XM_017009814.1 | XP_016865303.1 | 8 - 9 | 1450 | 97 | |
XM_017009815.1 | XP_016865304.1 | 8 - 9 | 1450 | 97 | |
XM_017009817.1 | XP_016865306.1 | 4 - 5 | 844 | 97 | |
GenBank mRNA | AK307154.1 | - | 3 - 4 | 553 | 97 |