The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100176.1 | NP_001093646.1 | 20 - 21 | 2107 | 67 |
NM_013312.2 | NP_037444.2 | 21 - 22 | 2113 | 67 | |
XM_005259873.2 | XP_005259930.1 | 21 - 22 | 2110 | 67 | |
XM_005259874.2 | XP_005259931.1 | 20 - 21 | 2104 | 67 | |
XM_005259876.4 | XP_005259933.1 | 21 - 22 | 2109 | 67 | |
XM_005259877.4 | XP_005259934.1 | 21 - 22 | 2059 | 67 | |
XM_011527943.2 | XP_011526245.1 | 21 - 22 | 3095 | 67 | |
XM_011527944.2 | XP_011526246.1 | 21 - 22 | 3094 | 67 | |
XM_011527945.2 | XP_011526247.1 | 21 - 22 | 3061 | 67 | |
XM_011527946.2 | XP_011526248.1 | 21 - 22 | 3086 | 67 | |
XM_011527947.2 | XP_011526249.1 | 17 - 18 | 1664 | 67 | |
XM_017026720.1 | XP_016882209.1 | 21 - 22 | 2083 | 67 | |
XM_017026721.1 | XP_016882210.1 | 20 - 21 | 2103 | 67 | |
GenBank mRNA | AF044924.1 | - | 21 - 22 | 2016 | 67 |
BC012443.1 | - | 20 - 21 | 2030 | 67 |