The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100176.1 | NP_001093646.1 | 21 - 22 | 2179 | 99 |
NM_013312.2 | NP_037444.2 | 22 - 23 | 2185 | 99 | |
XM_005259873.2 | XP_005259930.1 | 22 - 23 | 2182 | 99 | |
XM_005259874.2 | XP_005259931.1 | 21 - 22 | 2176 | 99 | |
XM_005259876.4 | XP_005259933.1 | 22 - 23 | 2181 | 99 | |
XM_005259877.4 | XP_005259934.1 | 22 - 23 | 2131 | 99 | |
XM_011527943.2 | XP_011526245.1 | 22 - 23 | 3167 | 99 | |
XM_011527944.2 | XP_011526246.1 | 22 - 23 | 3166 | 99 | |
XM_011527945.2 | XP_011526247.1 | 22 - 23 | 3133 | 99 | |
XM_011527946.2 | XP_011526248.1 | 22 - 23 | 3158 | 99 | |
XM_011527947.2 | XP_011526249.1 | 18 - 19 | 1736 | 99 | |
XM_017026720.1 | XP_016882209.1 | 22 - 23 | 2155 | 99 | |
XM_017026721.1 | XP_016882210.1 | 21 - 22 | 2175 | 99 | |
GenBank mRNA | AF044924.1 | - | 22 - 23 | 2088 | 99 |
BC012443.1 | - | 21 - 22 | 2102 | 99 |