The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015885.3 | NP_056969.2 | 11 - 12 | 4324 | 142 |
XM_005274048.1 | XP_005274105.1 | 11 - 12 | 4717 | 142 | |
XM_005274049.1 | XP_005274106.1 | 11 - 12 | 4714 | 142 | |
XM_017017918.1 | XP_016873407.1 | 11 - 12 | 4321 | 142 | |
GenBank mRNA | AB020631.1 | - | 11 - 12 | 4248 | 142 |
BC065384.1 | - | 11 - 12 | 4216 | 142 | |
BC146778.1 | - | 11 - 12 | 4248 | 142 |