The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015885.3 | NP_056969.2 | 15 - 16 | 4800 | 123 |
XM_005274048.1 | XP_005274105.1 | 15 - 16 | 5193 | 123 | |
XM_005274049.1 | XP_005274106.1 | 15 - 16 | 5190 | 123 | |
XM_017017918.1 | XP_016873407.1 | 15 - 16 | 4797 | 123 | |
GenBank mRNA | AB020631.1 | - | 15 - 16 | 4724 | 123 |
BC065384.1 | - | 15 - 16 | 4692 | 123 | |
BC146778.1 | - | 15 - 16 | 4724 | 123 |