Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OLFML2B
Entrez Gene ID
25903
Gene Name
olfactomedin like 2B
Gene Aliases
-
Chromosome Location
Chr.1: 161983192 - 162024465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297713.1 NP_001284642.1 2 - 3 1467 91
NM_015441.2 NP_056256.1 2 - 3 1467 91
XM_005245075.2 XP_005245132.1 2 - 3 1467 91
XM_017000967.1 XP_016856456.1 2 - 3 1467 91
GenBank mRNA AB119054.1 - 2 - 3 432 91
AK301499.1 - 2 - 3 642 91
AK316154.1 - 2 - 3 799 91
BC067274.1 - 2 - 3 856 91
BX648975.1 - 2 - 3 643 91

View Full Product Details