The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297713.1 | NP_001284642.1 | 5 - 6 | 1987 | 111 |
NM_015441.2 | NP_056256.1 | 5 - 6 | 1984 | 111 | |
XM_005245075.2 | XP_005245132.1 | 5 - 6 | 1990 | 111 | |
XM_011509398.2 | XP_011507700.1 | 2 - 3 | 2145 | 111 | |
XM_017000967.1 | XP_016856456.1 | 5 - 6 | 1990 | 111 | |
GenBank mRNA | AB119054.1 | - | 5 - 6 | 949 | 111 |
AK301499.1 | - | 5 - 6 | 1162 | 111 | |
AK316154.1 | - | 5 - 6 | 1316 | 111 | |
BC067274.1 | - | 5 - 6 | 1373 | 111 | |
BX648975.1 | - | 5 - 6 | 1160 | 111 |