The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284274.1 | NP_001271203.1 | 18 - 19 | 2002 | 143 |
NM_182765.4 | NP_877497.3 | 17 - 18 | 1990 | 143 | |
NR_104291.1 | 17 - 18 | 1961 | 143 | ||
XM_017015750.1 | XP_016871239.1 | 17 - 18 | 1840 | 143 | |
XM_017015752.1 | XP_016871241.1 | 10 - 11 | 1194 | 143 | |
GenBank mRNA | AK122629.1 | - | 12 - 13 | 1184 | 143 |
AK295723.1 | - | 18 - 19 | 1945 | 143 | |
AL713675.1 | - | 3 - 4 | 224 | 143 | |
BC040187.1 | - | 17 - 18 | 1933 | 143 |