Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HECTD2
Entrez Gene ID
143279
Gene Name
HECT domain E3 ubiquitin protein ligase 2
Gene Aliases
-
Chromosome Location
Chr.10: 91409280 - 91514829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284274.1 NP_001271203.1 19 - 20 2114 81
NM_182765.4 NP_877497.3 18 - 19 2102 81
NR_104291.1 18 - 19 2073 81
XM_017015750.1 XP_016871239.1 18 - 19 1952 81
XM_017015752.1 XP_016871241.1 11 - 12 1306 81
GenBank mRNA AK122629.1 - 13 - 14 1296 81
AK295723.1 - 19 - 20 2057 81
AL713675.1 - 4 - 5 336 81
BC040187.1 - 18 - 19 2045 81

View Full Product Details