Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTPBP3
Entrez Gene ID
84705
Gene Name
GTP binding protein 3 (mitochondrial)
Gene Aliases
COXPD23, GTPBG3, MSS1, MTGP1, THDF1
Chromosome Location
Chr.19: 17334982 - 17342731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128855.2 NP_001122327.1 6 - 7 900 135
NM_001195422.1 NP_001182351.1 6 - 7 893 135
NM_032620.3 NP_116009.2 6 - 7 900 135
NM_133644.3 NP_598399.2 5 - 6 996 135
GenBank mRNA AF360742.1 - 6 - 7 862 135
AF360743.1 - 6 - 7 862 135
AF360744.1 - 6 - 7 842 135
AK027606.1 - 6 - 7 847 135
AK091035.1 - 5 - 6 1701 135
AK291929.1 - 6 - 7 844 135
AK297953.1 - 6 - 7 875 135
AK298485.1 - - 557 135
AK311442.1 - 6 - 7 963 135
AY078987.1 - 5 - 6 958 135
AY078988.1 - 6 - 7 862 135
BC008924.2 - 4 - 5 440 135
BC017207.2 - 6 - 7 832 135
BC019261.1 - 6 - 7 841 135
BM789389.1 - 3 - 4 364 135

View Full Product Details