The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012474.4 | NP_036606.2 | 2 - 3 | 597 | 114 |
XM_017002236.1 | XP_016857725.1 | 3 - 4 | 802 | 114 | |
GenBank mRNA | AB062451.1 | - | 2 - 3 | 216 | 114 |
AF236637.1 | - | 2 - 3 | 276 | 114 | |
AK294537.1 | - | 2 - 3 | 270 | 114 | |
BC002906.2 | - | 2 - 3 | 437 | 114 | |
DR762696.1 | - | 2 - 3 | 597 | 114 |