The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001302623.1 | NP_001289552.1 | 13 - 14 | 2044 | 67 |
NM_001322216.1 | NP_001309145.1 | 13 - 14 | 2178 | 67 | |
NM_001322217.1 | NP_001309146.1 | 12 - 13 | 2010 | 67 | |
NM_001322218.1 | NP_001309147.1 | 13 - 14 | 2090 | 67 | |
NM_001322219.1 | NP_001309148.1 | 13 - 14 | 2096 | 67 | |
NM_001322220.1 | NP_001309149.1 | 13 - 14 | 2096 | 67 | |
NM_001322221.1 | NP_001309150.1 | 13 - 14 | 2090 | 67 | |
NM_004941.2 | NP_004932.1 | 13 - 14 | 2044 | 67 | |
NR_136225.1 | 13 - 14 | 2115 | 67 | ||
NR_136226.1 | 13 - 14 | 2096 | 67 | ||
NR_136227.1 | 13 - 14 | 2096 | 67 | ||
NR_136228.1 | 13 - 14 | 2096 | 67 | ||
GenBank mRNA | AK291664.1 | - | 13 - 14 | 1977 | 67 |
AK294046.1 | - | 12 - 13 | 2010 | 67 | |
AK303319.1 | - | 13 - 14 | 1998 | 67 | |
BC044586.1 | - | 13 - 14 | 1942 | 67 | |
BC047327.1 | - | 13 - 14 | 1946 | 67 | |
D50487.1 | - | 13 - 14 | 1996 | 67 |