The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001302623.1 | NP_001289552.1 | 7 - 8 | 1127 | 104 |
NM_001322216.1 | NP_001309145.1 | 7 - 8 | 1261 | 104 | |
NM_001322217.1 | NP_001309146.1 | 6 - 7 | 1093 | 104 | |
NM_001322218.1 | NP_001309147.1 | 7 - 8 | 1173 | 104 | |
NM_001322219.1 | NP_001309148.1 | 7 - 8 | 1179 | 104 | |
NM_001322220.1 | NP_001309149.1 | 7 - 8 | 1179 | 104 | |
NM_001322221.1 | NP_001309150.1 | 7 - 8 | 1173 | 104 | |
NM_004941.2 | NP_004932.1 | 7 - 8 | 1127 | 104 | |
NR_136225.1 | 7 - 8 | 1198 | 104 | ||
NR_136226.1 | 7 - 8 | 1179 | 104 | ||
NR_136227.1 | 7 - 8 | 1179 | 104 | ||
NR_136228.1 | 7 - 8 | 1179 | 104 | ||
GenBank mRNA | AK291664.1 | - | 7 - 8 | 1060 | 104 |
AK294046.1 | - | 6 - 7 | 1093 | 104 | |
AK303319.1 | - | 7 - 8 | 1081 | 104 | |
BC044586.1 | - | 7 - 8 | 1025 | 104 | |
BC047327.1 | - | 7 - 8 | 1029 | 104 | |
D50487.1 | - | 7 - 8 | 1079 | 104 |