The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320905.1 | NP_001307834.1 | 4 - 5 | 635 | 72 |
NM_012145.3 | NP_036277.2 | 3 - 4 | 518 | 72 | |
NR_135492.1 | 3 - 4 | 480 | 72 | ||
GenBank mRNA | AK223434.1 | - | 3 - 4 | 365 | 72 |
BC001827.2 | - | 3 - 4 | 72 | ||
BC147006.1 | - | 3 - 4 | 457 | 72 | |
BC147010.1 | - | 3 - 4 | 457 | 72 | |
BC171894.1 | - | 3 - 4 | 72 | ||
BF951595.1 | - | 2 - 3 | 72 | ||
BM552707.1 | - | 3 - 4 | 504 | 72 | |
CR541986.1 | - | 3 - 4 | 333 | 72 | |
CR542015.1 | - | 3 - 4 | 333 | 72 | |
CX166352.1 | - | 3 - 4 | 72 | ||
L16991.1 | - | 3 - 4 | 72 | ||
X54729.1 | - | 3 - 4 | 72 |