Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BSPRY
Entrez Gene ID
54836
Gene Name
B-box and SPRY domain containing
Gene Aliases
-
Chromosome Location
Chr.9: 113349532 - 113371233 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317943.1 NP_001304872.1 2 - 3 353 103
NM_001317944.1 NP_001304873.1 2 - 3 353 103
NM_017688.2 NP_060158.2 2 - 3 353 103
XM_006717149.3 XP_006717212.1 2 - 3 344 103
GenBank mRNA AJ276691.1 - 2 - 3 334 103
AK092607.1 - 2 - 3 344 103
BC001477.2 - 2 - 3 362 103
BP370567.1 - 2 - 3 176 103

View Full Product Details