The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004463.2 | NP_004454.2 | 4 - 5 | 1839 | 79 |
GenBank mRNA | AK226117.1 | - | 4 - 5 | 1827 | 79 |
AK299635.1 | - | 2 - 3 | 548 | 79 | |
BC034530.1 | - | 4 - 5 | 1351 | 79 | |
U11690.1 | - | 4 - 5 | 1835 | 79 |