The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042601.2 | NP_001036066.1 | 5 - 6 | 832 | 137 |
NM_001288582.1 | NP_001275511.1 | 5 - 6 | 832 | 137 | |
NM_133462.3 | NP_597719.1 | 5 - 6 | 832 | 137 | |
XM_017005740.1 | XP_016861229.1 | 5 - 6 | 819 | 137 | |
GenBank mRNA | AB075860.2 | - | 3 - 4 | 370 | 137 |
AK055984.1 | - | 5 - 6 | 790 | 137 | |
AL832052.1 | - | 5 - 6 | 770 | 137 | |
AL833086.2 | - | 5 - 6 | 769 | 137 | |
AY358762.1 | - | 5 - 6 | 752 | 137 | |
BC027990.1 | - | 5 - 6 | 765 | 137 |